Learn More
TARGETMOL CHEMICALS INC Cefotiam hydrochloride 200MG
SDP
Catalog No. 502877922
Encompass_Preferred
Also available in 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Cefotiam hydrochloride (SCE-963 hydrochloride) is the hydrochloride salt form of cefotiam, a third-generation, semi-synthetic, beta-lactam cephalosporin antibiotic with antibacterial activity. Cefotiam binds to penicillin-binding proteins (PBPs), transpeptidases that are responsible for crosslinking of peptidoglycan. By preventing crosslinking of peptidoglycan, cell wall integrity is lost and cell wall synthesis is halted. Purity 98.74%
Catalog No. 50-287-7922 Supplier TARGETMOL CHEMICALS INC Supplier No. T1616200MG
Also available in 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Cefotiam hydrochloride (SCE-963 hydrochloride) is the hydrochloride salt form of cefotiam, a third-generation, semi-synthetic, beta-lactam cephalosporin antibiotic with antibacterial activity. Cefotiam binds to penicillin-binding proteins (PBPs), transpeptidases that are responsible for crosslinking of peptidoglycan. By preventing crosslinking of peptidoglycan, cell wall integrity is lost and cell wall synthesis is halted. Purity 98.74%