Learn More
Medchemexpress LLC Cefotiam hydrochloride | 66309-69-1 | 98.0% | 100 MG
SDP
Catalog No. 5000406876
Encompass_Preferred
Cefotiam hydrochloride is a parenteral cephalosporin antibiotic that exhibits broad-spectrum activity against both Gram-positive and Gram-negative bacteria. It targets β-lactam and is suitable for various research applications.
  • Exhibits antibacterial activity with a MIC value of 1.56 μg/mL for *P. mirabilis* IFO 3849.
  • Highly active against *Staph. aureus* (MIC values of 0.5-1 μg/mL) and *Staph. albus* (MICs 0.25-0.5 μg/mL).
  • Effective against haemolytic streptococci, pneumococci, and *Streptococcus viridans* with MIC values in the range of 0.06-4 μg/mL.
  • Demonstrates in vivo efficacy in curing urinary tract infection with *P. mirabilis* in mice.
Catalog No. 50-004-06876 Supplier Medchemexpress LLC Supplier No. HYB0734A100MG
Cefotiam hydrochloride is a parenteral cephalosporin antibiotic that exhibits broad-spectrum activity against both Gram-positive and Gram-negative bacteria. It targets β-lactam and is suitable for various research applications.
  • Exhibits antibacterial activity with a MIC value of 1.56 μg/mL for *P. mirabilis* IFO 3849.
  • Highly active against *Staph. aureus* (MIC values of 0.5-1 μg/mL) and *Staph. albus* (MICs 0.25-0.5 μg/mL).
  • Effective against haemolytic streptococci, pneumococci, and *Streptococcus viridans* with MIC values in the range of 0.06-4 μg/mL.
  • Demonstrates in vivo efficacy in curing urinary tract infection with *P. mirabilis* in mice.