Learn More
Medchemexpress LLC Cefotiam hydrochloride | 66309-69-1 | 98.0% | 50 MG
SDP
Supplier:  Medchemexpress LLC HYB0734A50MG
Encompass_Preferred
Cefotiam hydrochloride is a parenteral cephalosporin antibiotic known for its broad-spectrum activity against both Gram-positive and Gram-negative bacteria.
  • Parenteral cephalosporin antibiotic
  • Broad-spectrum activity
  • Effective against Gram-positive bacteria
  • Effective against Gram-negative bacteria
Catalog No. 50-004-06759