Learn More
Cayman Chemical Cefotiam (hydrochloride hydrate), Purity: ≥80%, 5 g
SDP
Catalog No. 502948639
Encompass_Preferred
A broad-spectrum cephalosporin antibiotic; inhibits the growth of S. aureus (MIC50 = 0.26 µg/ml), as well as P. aeruginosa, E. coli, and H. influenzae (MIC50s = 1, 0.06, and 0.01 µg/ml, respectively); increases survival in mouse models of infection with strains of S. aureus (ED50s = 3.5 and 8.3 mg/kg) and E. coli (ED50s = 0.5-25 mg/kg)
Catalog No. 50-294-8639 Supplier Cayman Chemical Supplier No. 328715 G
A broad-spectrum cephalosporin antibiotic; inhibits the growth of S. aureus (MIC50 = 0.26 µg/ml), as well as P. aeruginosa, E. coli, and H. influenzae (MIC50s = 1, 0.06, and 0.01 µg/ml, respectively); increases survival in mouse models of infection with strains of S. aureus (ED50s = 3.5 and 8.3 mg/kg) and E. coli (ED50s = 0.5-25 mg/kg)