Learn More
Medchemexpress LLC HY-B1825 100mg Medchemexpress, Cefoxitin CAS:35607-66-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1825100MG
Encompass_Preferred
Medchemexpress, HY-B1825 100mg Cefoxitin CAS:35607-66-0 Cefoxitin is a broad-spectrum, second-generation cephalosporin with antibacterial activity. Cefoxitin is effective against a wide variety of infections caused by gram-positive or gram-negative aerobes as well as by anaerobic bacteria[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1233