Learn More
Medchemexpress LLC Cefoxitin | 35607-66-0 | 99.9% | 250 MG
SDP
Catalog No. 5000414435
Encompass_Preferred
Cefoxitin is a cephalosporin-class antibiotic with a broad spectrum of antibacterial activity, effective against both Gram-negative and Gram-positive bacteria. It exhibits high stability against β-Lactamase.
  • Highly stable against β-Lactamase
  • Broad spectrum of antibacterial activity
  • Effective against Gram-negative bacteria
  • Effective against Gram-positive bacteria
Catalog No. 50-004-14435 Supplier Medchemexpress LLC Supplier No. 250MGHYB1825
Cefoxitin is a cephalosporin-class antibiotic with a broad spectrum of antibacterial activity, effective against both Gram-negative and Gram-positive bacteria. It exhibits high stability against β-Lactamase.
  • Highly stable against β-Lactamase
  • Broad spectrum of antibacterial activity
  • Effective against Gram-negative bacteria
  • Effective against Gram-positive bacteria