Learn More
Medchemexpress LLC Cefoxitin (sodium) | 33564-30-6 | 98.8% | 250 MG
SDP
Catalog No. 5000414325
Encompass_Preferred
Cefoxitin (sodium) (Standard) is a light yellow to yellow solid with a molecular weight of 449.43 and molecular formula C₁₆H₁₆N₃NaO₇S₂. It has a purity of 98.82% as determined by HPLC.
  • Consistent with structure by 1H NMR and MS.
  • Purity not less than 90.0%.
  • Traceability to ISO 17025 requirements.
  • ISO17025, ISO9001, and GMP certified.
Catalog No. 50-004-14325 Supplier Medchemexpress LLC Supplier No. 250MGHYB1117R
Cefoxitin (sodium) (Standard) is a light yellow to yellow solid with a molecular weight of 449.43 and molecular formula C₁₆H₁₆N₃NaO₇S₂. It has a purity of 98.82% as determined by HPLC.
  • Consistent with structure by 1H NMR and MS.
  • Purity not less than 90.0%.
  • Traceability to ISO 17025 requirements.
  • ISO17025, ISO9001, and GMP certified.