Learn More
Apexbio Technology LLC Cefoxitin (sodium salt) 33564-30-6 500mg
SDP
Catalog No. 5000091608
Encompass_Preferred
Cefoxitin sodium salt (CAS 33564-30-6) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit PBPs thereby interfering with bacterial cell wall synthesis and peptidoglycan cross-linking Cefoxitin sodium salt exerts its biological activity primarily through inhibition of cell wall synthesis by binding to PBPs In in vitro enzymatic assays Cefoxitin sodium salt demonstrates inhibitory activity against various gram-positive and gram-negative bacteria with previously reported IC50 values in the low micromolar range due to its resistance to certain beta-lactamases including AmpC -lactamases Based on these pharmacological properties Cefoxitin sodium salt holds research potential in investigations of bacterial resistance mechanisms antimicrobial sensitivity profiling and beta-lactamase enzyme kinetics
Catalog No. 50-000-91608 Supplier Apexbio Technology LLC Supplier No. C4849500
Cefoxitin sodium salt (CAS 33564-30-6) is a small-molecule inhibitor targeting penicillin-binding proteins (PBPs) It is designed to inhibit PBPs thereby interfering with bacterial cell wall synthesis and peptidoglycan cross-linking Cefoxitin sodium salt exerts its biological activity primarily through inhibition of cell wall synthesis by binding to PBPs In in vitro enzymatic assays Cefoxitin sodium salt demonstrates inhibitory activity against various gram-positive and gram-negative bacteria with previously reported IC50 values in the low micromolar range due to its resistance to certain beta-lactamases including AmpC -lactamases Based on these pharmacological properties Cefoxitin sodium salt holds research potential in investigations of bacterial resistance mechanisms antimicrobial sensitivity profiling and beta-lactamase enzyme kinetics