Learn More
Medchemexpress LLC HY-B0771A 50mg Medchemexpress, Cefozopran (hydrochloride) CAS:113981-44-5 Purity:>98%
SDP
Catalog No. 502082661
Encompass_Preferred
Medchemexpress, HY-B0771A 50mg Cefozopran (hydrochloride) CAS:113981-44-5 Cefozopran (SCE-2787) hydrochloride is a semi-synthetic, parenteral, fourth-generation cephalosporin. Cefozopran hydrochloride, an antibiotic, has a broad spectrum of antibacterial activity, inhibiting most of the gram-negative and gram-positive organisms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2661 Supplier Medchemexpress LLC Supplier No. HYB0771A50MG
Medchemexpress, HY-B0771A 50mg Cefozopran (hydrochloride) CAS:113981-44-5 Cefozopran (SCE-2787) hydrochloride is a semi-synthetic, parenteral, fourth-generation cephalosporin. Cefozopran hydrochloride, an antibiotic, has a broad spectrum of antibacterial activity, inhibiting most of the gram-negative and gram-positive organisms. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.