Learn More
Medchemexpress LLC Cefozopran hydrochloride | 113981-44-5 | 95.0% | 100 MG
SDP
Catalog No. 5000408111
Encompass_Preferred
Cefozopran hydrochloride is a semi-synthetic, parenteral, fourth-generation cephalosporin. This antibiotic exhibits a broad spectrum of antibacterial activity, effectively inhibiting most gram-negative and gram-positive organisms.
  • Exhibits broad-spectrum antibacterial activity.
  • Effectively inhibits most gram-negative and gram-positive organisms.
  • Shows good activity against methicillin-susceptible staphylococci, enterococci, and viridans group streptococci.
  • Active against gram-negative organisms, including Haemophilus influenza.
  • Demonstrates comparatively good activity against enterococci and P. aeruginosa.
  • Effective against acute respiratory tract infections in mice models.
  • Effective against chronic respiratory tract infection in mice models.
Catalog No. 50-004-08111 Supplier Medchemexpress LLC Supplier No. HYB0771A100MG
Cefozopran hydrochloride is a semi-synthetic, parenteral, fourth-generation cephalosporin. This antibiotic exhibits a broad spectrum of antibacterial activity, effectively inhibiting most gram-negative and gram-positive organisms.
  • Exhibits broad-spectrum antibacterial activity.
  • Effectively inhibits most gram-negative and gram-positive organisms.
  • Shows good activity against methicillin-susceptible staphylococci, enterococci, and viridans group streptococci.
  • Active against gram-negative organisms, including Haemophilus influenza.
  • Demonstrates comparatively good activity against enterococci and P. aeruginosa.
  • Effective against acute respiratory tract infections in mice models.
  • Effective against chronic respiratory tract infection in mice models.