Learn More
Medchemexpress LLC Cefozopran hydrochloride | 113981-44-5 | MFCD00944908 | >95.0% | 1 ML
SDP
Catalog No. 5000409203
Encompass_Preferred
Cefozopran hydrochloride (SCE-2787) is a semi-synthetic, parenteral, fourth-generation cephalosporin antibiotic. This antibiotic demonstrates a broad spectrum of antibacterial activity, effectively inhibiting most gram-negative and gram-positive organisms.
  • Semi-synthetic
  • Parenteral, fourth-generation cephalosporin
  • Broad-spectrum antibacterial activity
  • Effectively inhibits most gram-negative organisms
  • Effectively inhibits most gram-positive organisms
Catalog No. 50-004-09203 Supplier Medchemexpress LLC Supplier No. HYB0771A10MM/1ML
Cefozopran hydrochloride (SCE-2787) is a semi-synthetic, parenteral, fourth-generation cephalosporin antibiotic. This antibiotic demonstrates a broad spectrum of antibacterial activity, effectively inhibiting most gram-negative and gram-positive organisms.
  • Semi-synthetic
  • Parenteral, fourth-generation cephalosporin
  • Broad-spectrum antibacterial activity
  • Effectively inhibits most gram-negative organisms
  • Effectively inhibits most gram-positive organisms