Learn More
Medchemexpress LLC Cefpiramide sodium | 74849-93-7 | 98.8% | 100 MG
SDP
Catalog No. 5000407529
Encompass_Preferred
Cefpiramide sodium is a Pseudomonas-active cephalosporin with a broad spectrum of antibacterial activity. It is intended for research use only.
  • Acts as an antibacterial agent
  • Moderately susceptible to hydrolysis by various beta-lactamases from Gram-negative bacilli
  • More active against Acinetobacter spp. and Pseudomonas spp.
  • Inhibits methicillin-susceptible staphylococci, non-enterococcal streptococci, Neisseria gonorrhoeae, N. meningitidis, and beta-lactamase-negative Haemophilus influenzae
  • Inhibited many Pseudomonas aeruginosa resistant to carbenicillin, piperacillin, and cefotaxime
  • Active against Streptococcus faecalis and Listeria moncytogenes
Catalog No. 50-004-07529 Supplier Medchemexpress LLC Supplier No. HYB0798100MG
Cefpiramide sodium is a Pseudomonas-active cephalosporin with a broad spectrum of antibacterial activity. It is intended for research use only.
  • Acts as an antibacterial agent
  • Moderately susceptible to hydrolysis by various beta-lactamases from Gram-negative bacilli
  • More active against Acinetobacter spp. and Pseudomonas spp.
  • Inhibits methicillin-susceptible staphylococci, non-enterococcal streptococci, Neisseria gonorrhoeae, N. meningitidis, and beta-lactamase-negative Haemophilus influenzae
  • Inhibited many Pseudomonas aeruginosa resistant to carbenicillin, piperacillin, and cefotaxime
  • Active against Streptococcus faecalis and Listeria moncytogenes