Learn More
Selleck Chemical LLC Cefpirome sulfate S3637-25mg
SDP
Catalog No. 503109266
Encompass_Preferred
Cefpirome(HR-810 sulfate) is a fourth-generation cephalosporin and is considered highly active against Gram-negative bacteria
Catalog No. 50-310-9266 Supplier Selleck Chemical LLC Supplier No. S363725MG
Cefpirome(HR-810 sulfate) is a fourth-generation cephalosporin and is considered highly active against Gram-negative bacteria