Learn More
Medchemexpress LLC Cefpirome sulfate | 98753-19-6 | 99.5% | 250 MG
SDP
Catalog No. 5000414495
Encompass_Preferred
Cefpirome sulfate is a cephalosporin antibiotic that effectively crosses cell membranes and the blood-brain barrier. It functions by binding with high affinity to penicillin-binding proteins, thereby inhibiting the synthesis of bacterial cell walls. This antibiotic demonstrates both bactericidal and growth-inhibitory activities against various Gram-negative bacteria, Gram-positive bacteria, and susceptible anaerobic bacteria, including certain β-lactamase-producing strains. It is intended strictly for research purposes.
  • Inhibits bacterial cell wall synthesis
  • Exhibits bactericidal and growth-inhibitory activities
  • Active against Gram-negative and Gram-positive bacteria
  • Effective against susceptible anaerobic bacteria
  • Active against some β-lactamase-producing strains
Catalog No. 50-004-14495 Supplier Medchemexpress LLC Supplier No. 250MGHYB1824
Cefpirome sulfate is a cephalosporin antibiotic that effectively crosses cell membranes and the blood-brain barrier. It functions by binding with high affinity to penicillin-binding proteins, thereby inhibiting the synthesis of bacterial cell walls. This antibiotic demonstrates both bactericidal and growth-inhibitory activities against various Gram-negative bacteria, Gram-positive bacteria, and susceptible anaerobic bacteria, including certain β-lactamase-producing strains. It is intended strictly for research purposes.
  • Inhibits bacterial cell wall synthesis
  • Exhibits bactericidal and growth-inhibitory activities
  • Active against Gram-negative and Gram-positive bacteria
  • Effective against susceptible anaerobic bacteria
  • Active against some β-lactamase-producing strains