Learn More
Medchemexpress LLC Cefpirome sulfate | 98753-19-6 | MFCD08234364 | 99.0% | 612.66 g/mol | C22H24N6O9S3 | 10 MG
SDP
Catalog No. 5000298030
Encompass_Preferred
Cefpirome sulfate is an analytical reference material of cefpirome, a fourth-generation cephalosporin antibiotic. The substance is supplied as a light yellow to yellow solid with identity confirmed by 1H NMR and mass spectrometry, and accompanied by a certificate of analysis reporting HPLC purity and assay results. Stored sealed and dry at 4°C, it is intended for research and analytical applications.
Catalog No. 50-002-98030 Supplier Medchemexpress LLC Supplier No. HYB1824R10MG
Cefpirome sulfate is an analytical reference material of cefpirome, a fourth-generation cephalosporin antibiotic. The substance is supplied as a light yellow to yellow solid with identity confirmed by 1H NMR and mass spectrometry, and accompanied by a certificate of analysis reporting HPLC purity and assay results. Stored sealed and dry at 4°C, it is intended for research and analytical applications.