Learn More
Cayman Chemical Cefpodoxime, Purity: ≥95%, CAS: 80210-62-4, 100 mg
SDP
Catalog No. 502971660
Encompass_Preferred
A cephalosporin antibiotic; a metabolite of cefpodoxime proxetil; active against both Gram-positive and Gram-negative bacteria (MIC90s = 0.015-8 µg/ml)
Catalog No. 50-297-1660 Supplier Cayman Chemical Supplier No. 16075100 MG
A cephalosporin antibiotic; a metabolite of cefpodoxime proxetil; active against both Gram-positive and Gram-negative bacteria (MIC90s = 0.015-8 µg/ml)