Learn More
AdipoGen Cefpodoxime
SDP
Catalog No. 502561364
Encompass_Preferred
Chemical. CAS 80210-62-4. Formula C15H17N5O6S2. MW 427.46. Synthetic. Cefpodoxime is a metabolite of cefpodoxime proxetil, an oral third generation cephalosporin antibiotic. It is active against most Gram positive and Gram negative bacteria MIC90 ranges from 0.015-8 µg/ml. It is commonly used to treat acute otitis media, pharyngitis, and sinusitis. Cefpodoxime inhibits cell wall synthesis by inhibiting the final transpeptidation step of peptidoglycan synthesis in cell walls.
Catalog No. 50-256-1364 Supplier AdipoGen Supplier No. CDXC0361M100
Chemical. CAS 80210-62-4. Formula C15H17N5O6S2. MW 427.46. Synthetic. Cefpodoxime is a metabolite of cefpodoxime proxetil, an oral third generation cephalosporin antibiotic. It is active against most Gram positive and Gram negative bacteria MIC90 ranges from 0.015-8 µg/ml. It is commonly used to treat acute otitis media, pharyngitis, and sinusitis. Cefpodoxime inhibits cell wall synthesis by inhibiting the final transpeptidation step of peptidoglycan synthesis in cell walls.