Learn More
Cayman Chemical Cefpodoxime, Purity: ≥95%, CAS: 80210-62-4, 25 mg
SDP
Catalog No. 502972944
Encompass_Preferred
A cephalosporin antibiotic; a metabolite of cefpodoxime proxetil; active against both Gram-positive and Gram-negative bacteria (MIC90s = 0.015-8 µg/ml)
Catalog No. 50-297-2944 Supplier Cayman Chemical Supplier No. 1607525 MG
A cephalosporin antibiotic; a metabolite of cefpodoxime proxetil; active against both Gram-positive and Gram-negative bacteria (MIC90s = 0.015-8 µg/ml)