Learn More
Cayman Chemical CefpodoxIme 50mg
SDP
Catalog No. 502972945
Encompass_Preferred
A cephalosporin antibiotic; a metabolite of cefpodoxime proxetil; active against both Gram-positive and Gram-negative bacteria (MIC90s = 0.015-8 UG/ml)
Catalog No. 50-297-2945 Supplier Cayman Chemical Supplier No. 1607550 MG
A cephalosporin antibiotic; a metabolite of cefpodoxime proxetil; active against both Gram-positive and Gram-negative bacteria (MIC90s = 0.015-8 UG/ml)