Learn More
Medchemexpress LLC HY-N7101 25mg Medchemexpress, Cefpodoxime Proxetil CAS:87239-81-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN710125MG
Encompass_Preferred
Medchemexpress, HY-N7101 25mg Cefpodoxime Proxetil CAS:87239-81-4 Cefpodoxime Proxetil is a first oral and broad spectrum antibiotic that belongs to the third generation of cephalosporin. Cefpodoxime Proxetil binds to penicillin binding proteins (PBPs), which inhibits peptidoglycan synthesis, finally results in interfering bacterial cell wall biosynthesis. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-1612