Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefpodoxime Sodium Salt, G-Biosciences
SCI_ED

Catalog No. S902523
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S902523 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S902523 Supplier G-Biosciences Supplier No. RC939
Add to Cart
Add to Cart

Cefpodoxime sodium is a third generation cephalosporin antibiotic.

It targets gram-positive and gram-negative bacteria, especially those responsible for otitis media and pharyngitis.

Chemical Identifiers

Molecular Weight (g/mol) 449.44 g/mol

Specifications

Boiling Point 200 to 202°C
Quantity 100 mg
Percent Purity >98%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.