Learn More
Apexbio Technology LLC Cefprozil 92665-29-7 5mg
SDP
Catalog No. 502240570
Encompass_Preferred
Cefprozil (CAS 92665-29-7) is a second-generation semisynthetic cephalosporin antibiotic targeting penicillin-binding proteins (PBPs) It is designed to resist hydrolysis by bacterial -lactamases via its molecular structure thereby inhibiting peptidoglycan synthesis in the bacterial cell wall Cefprozil exerts its biological activity primarily through selective binding to PBPs leading to inhibition of cell wall synthesis and subsequent bacterial lysis Reported IC50 values vary depending on bacterial species and experimental conditions Based on these pharmacological properties Cefprozil holds research potential in the investigation of bacterial resistance mechanisms evaluation of bacterial response and resistance-related gene expression and pharmacokinetic and pharmacodynamic studies of cephalosporin antibiotics
This item is not returnable. View return policy
Catalog No. 50-224-0570 Supplier Apexbio Technology LLC Supplier No. B49745
Cefprozil (CAS 92665-29-7) is a second-generation semisynthetic cephalosporin antibiotic targeting penicillin-binding proteins (PBPs) It is designed to resist hydrolysis by bacterial -lactamases via its molecular structure thereby inhibiting peptidoglycan synthesis in the bacterial cell wall Cefprozil exerts its biological activity primarily through selective binding to PBPs leading to inhibition of cell wall synthesis and subsequent bacterial lysis Reported IC50 values vary depending on bacterial species and experimental conditions Based on these pharmacological properties Cefprozil holds research potential in the investigation of bacterial resistance mechanisms evaluation of bacterial response and resistance-related gene expression and pharmacokinetic and pharmacodynamic studies of cephalosporin antibiotics