Learn More
Cayman Chemical Cefprozil-d4, Purity: ≥99% deuterated forms (d1-d4), CAS: 1426173-48-9, 1 mg
SDP
Catalog No. 502948032
Encompass_Preferred
An internal standard for the quantification of cefprozil by GC- or LC-MS
Catalog No. 50-294-8032 Supplier Cayman Chemical Supplier No. 336391 MG
An internal standard for the quantification of cefprozil by GC- or LC-MS