Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefprozil, G-Biosciences
SCI_ED

Catalog No. S787101
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S787101 50 mg
S000012 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S787101 Supplier G-Biosciences Supplier No. RC942
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefprozil is a second generation cephalosporin antibiotic targeting gram-negative and gram-positive bacteria. It shows some resistance to Beta-lactamase activity.

It works by stopping the growth of bacteria.

Chemical Identifiers

Molecular Weight (g/mol) 389.43

Specifications

Physical Form Powder
Quantity 50 mg
Product Line Gibco
Boiling Point 803°C
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.