Learn More
Apexbio Technology LLC Cefprozil hydrate 121123-17-9 5mg
SDP
Catalog No. 502240595
Encompass_Preferred
Cefprozil hydrate is a small-molecule inhibitor targeting bacterial penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis by blocking PBPs thereby disrupting peptidoglycan cross-linking and compromising cell wall integrity Cefprozil hydrate exerts its biological activity primarily through inhibition of PBPs leading to bacterial cell lysis and death In in vitro studies Cefprozil hydrate demonstrates antibacterial activity against both Gram-positive and Gram-negative microorganisms Based on these pharmacological properties Cefprozil hydrate holds research potential in antibacterial spectrum characterization bacterial resistance profiling comparator or control use in susceptibility testing and evaluation of novel antibacterial drug candidates
This item is not returnable. View return policy
Catalog No. 50-224-0595 Supplier Apexbio Technology LLC Supplier No. B21425
Cefprozil hydrate is a small-molecule inhibitor targeting bacterial penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis by blocking PBPs thereby disrupting peptidoglycan cross-linking and compromising cell wall integrity Cefprozil hydrate exerts its biological activity primarily through inhibition of PBPs leading to bacterial cell lysis and death In in vitro studies Cefprozil hydrate demonstrates antibacterial activity against both Gram-positive and Gram-negative microorganisms Based on these pharmacological properties Cefprozil hydrate holds research potential in antibacterial spectrum characterization bacterial resistance profiling comparator or control use in susceptibility testing and evaluation of novel antibacterial drug candidates