Learn More
Selleck Chemical LLC Cefquinome sulfate S5332-25mg
SDP
Catalog No. 503109965
Encompass_Preferred
Cefquinome sulphate is a veterinary parenteral and fourth-generation cephalosporin Its antimicrobial potency and extensive antibacterial spectrum result from the introduction of a methoxyimino-aminothiazolyl moiety into the acyl side chain
Catalog No. 50-310-9965 Supplier Selleck Chemical LLC Supplier No. S533225MG
Cefquinome sulphate is a veterinary parenteral and fourth-generation cephalosporin Its antimicrobial potency and extensive antibacterial spectrum result from the introduction of a methoxyimino-aminothiazolyl moiety into the acyl side chain