Learn More
Medchemexpress LLC Cefquinome (sulfate) | 118443-89-3 | 1 ML
SDP
Supplier:  Medchemexpress LLC HYN666510MM/1ML
Encompass_Preferred
Cefquinome (sulfate) is a broad-spectrum cephem antibiotic that has inhibitory effects on a variety of Gram-positive and Gram-negative bacteria, including Staphylococci, Streptococci, Pseudomonas, and Enterobacteriaceae.
  • Inhibits a variety of Gram-positive and Gram-negative bacteria.
  • Broad-spectrum cephem antibiotic.
Catalog No. 50-004-17880