Learn More
Medchemexpress LLC Cefquinome sulfate | 118443-89-3 | 50 MG
SDP
Catalog No. 5000419965
Encompass_Preferred
Cefquinome sulfate is a broad-spectrum cephem antibiotic that inhibits various Gram-positive and Gram-negative bacteria, including Staphylococci, Streptococci, Pseudomonas, and Enterobacteriaceae. It functions by binding to penicillin-binding proteins (PBPs), thereby inhibiting the synthesis of bacterial cell walls.
  • Inhibits methicillin susceptible S. aureus, Streptococcus spp., Enterococci, E. coli, Salmonella spp., Klebsiella spp., S. marcescens, Enterobacter spp., Citrobacter spp., P. mirabilis, Indole-positive Proteus spp., and P. aeruginosa
  • Shows in vivo anti-infectious activity against Klebsiella pneumoniae in a rat pneumonia model
  • Reduces lung inflammatory cell infiltration and lung tissue damage
  • Inhibits expression of inflammatory factors (TNF-α, IL-1β, IL-8)
  • Promotes expression of anti-inflammatory factors (IL-4, IL-10, IL-13)
Catalog No. 50-004-19965 Supplier Medchemexpress LLC Supplier No. HYN666550MG
Cefquinome sulfate is a broad-spectrum cephem antibiotic that inhibits various Gram-positive and Gram-negative bacteria, including Staphylococci, Streptococci, Pseudomonas, and Enterobacteriaceae. It functions by binding to penicillin-binding proteins (PBPs), thereby inhibiting the synthesis of bacterial cell walls.
  • Inhibits methicillin susceptible S. aureus, Streptococcus spp., Enterococci, E. coli, Salmonella spp., Klebsiella spp., S. marcescens, Enterobacter spp., Citrobacter spp., P. mirabilis, Indole-positive Proteus spp., and P. aeruginosa
  • Shows in vivo anti-infectious activity against Klebsiella pneumoniae in a rat pneumonia model
  • Reduces lung inflammatory cell infiltration and lung tissue damage
  • Inhibits expression of inflammatory factors (TNF-α, IL-1β, IL-8)
  • Promotes expression of anti-inflammatory factors (IL-4, IL-10, IL-13)