Learn More
Cayman Chemical Cefquinome (sulfate), Purity: ≥98%, CAS: 118443-89-3, 250 mg
SDP
Catalog No. 502949680
Encompass_Preferred
A broad-spectrum cephalosporin antibiotic; active against clinical isolates of various Gram-positive and Gram-negative bacteria, Salmonella, E. coli, and MRSA (MICs = 0.06-0.5, 0.015-0.5, and 1-16 µg/ml, respectively); intramammary administration is curative in a bovine model of E. coli-induced mastitis at 75 mg/animal
Catalog No. 50-294-9680 Supplier Cayman Chemical Supplier No. 34266250 MG
A broad-spectrum cephalosporin antibiotic; active against clinical isolates of various Gram-positive and Gram-negative bacteria, Salmonella, E. coli, and MRSA (MICs = 0.06-0.5, 0.015-0.5, and 1-16 µg/ml, respectively); intramammary administration is curative in a bovine model of E. coli-induced mastitis at 75 mg/animal