Learn More
AG Scientific Inc Cefsulodin, Sodium Salt, 1 G
Catalog No. 502236905
Encompass_Preferred
Cefsulodin Sodium Salt is a third-generation cephalosporin antibiotic used in CIN Agar. It displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP). Cefsulodin is used to study the effect of expression, binding, and inhibition of PBP1 on bacterial cell wall mucopeptide synthesis.CAS Number: 52152-93-9Molecular Weight: 554.5Chemical Formula: C22H19N4NaO8S2Solubility: Freely soluble in water & formamide. Slightly soluble in methanol. Very slightly soluble in ethanol.Storage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.
Catalog No. 50-223-6905 Supplier AG Scientific Inc Supplier No. C14571GM
Cefsulodin Sodium Salt is a third-generation cephalosporin antibiotic used in CIN Agar. It displays a mechanism of action like many β lactam antibiotics through inhibition of cell wall synthesis by competitively inhibiting penicillin binding protein (PBP). Cefsulodin is used to study the effect of expression, binding, and inhibition of PBP1 on bacterial cell wall mucopeptide synthesis.CAS Number: 52152-93-9Molecular Weight: 554.5Chemical Formula: C22H19N4NaO8S2Solubility: Freely soluble in water & formamide. Slightly soluble in methanol. Very slightly soluble in ethanol.Storage Temperature: -20CResearch or further manufacturing use only, not for food or drug use.