Learn More
Apexbio Technology LLC CEFSULODIN-SODIUM-SALT-500MG
SDP
Catalog No. 5000568274
Encompass_Preferred
Cefsulodin (sodium salt) (CAS No 52152-93-9) is a -lactam antibiotic primarily used in microbiological research Originating from the cephalosporin family it functions by inhibiting bacterial cell wall synthesis targeting penicillin-binding proteins particularly effective against Pseudomonas aeruginosa Its in vitro activity is observed within the nanomolar to low micromolar range making it a valuable tool in evaluating bacterial susceptibility Cefsulodin s applications extend to various experimental setups including cell culture models animal studies and drug screening assays for antibacterial agents It serves as a benchmark in establishing the efficacy of new antimicrobial compounds Experimental concentrations typically depend on the specific research design allowing flexibility in its application to suit diverse investigative needs
Catalog No. 50-005-68274 Supplier Apexbio Technology LLC Supplier No. C5593500
Cefsulodin (sodium salt) (CAS No 52152-93-9) is a -lactam antibiotic primarily used in microbiological research Originating from the cephalosporin family it functions by inhibiting bacterial cell wall synthesis targeting penicillin-binding proteins particularly effective against Pseudomonas aeruginosa Its in vitro activity is observed within the nanomolar to low micromolar range making it a valuable tool in evaluating bacterial susceptibility Cefsulodin s applications extend to various experimental setups including cell culture models animal studies and drug screening assays for antibacterial agents It serves as a benchmark in establishing the efficacy of new antimicrobial compounds Experimental concentrations typically depend on the specific research design allowing flexibility in its application to suit diverse investigative needs