Learn More
Mabtech Inc PepPool: CEFTA (CD4), human (1 vial). 35 MHC class II peptides from CMV, EBV, influenza, tetanus toxin, and adenovirus 5. Positive control for human CD4 T-cell stimulation in ELISpot, FluoroSpot, and flow. >95% pure; 25 µg each peptide.
Catalog No. NC1925864
Encompass
PepPool: CEFTA (CD4), human
Peptide pool for use as a positive control for human CD4 T-cell stimulation in ELISpot, FluoroSpot, and flow cytometry. Consists of 35 MHC class II-restricted peptides from human CMV, EBV, influenza virus, tetanus toxin, and adenovirus 5. Designed for broad HLA coverage. Synthetic peptides >95% pure. Induces IFN-γ, IL-2, IL-5, and IL-13 from antigen-specific CD4 T cells. For research use only. Not for diagnostic use.
Contents: One vial, lyophilized peptide pool, 35 peptides, 25 µg of each.
HLA coverage: HLA-DRB1*01, *03, *04, *07, *08, *11, *13, *15, *16, *53; HLA-DPw2; HLA-DPB1*04; HLA-DQw1; HLA-DQw3; HLA-DQB1*06.
Benefits: Validated positive control for CD4 responses; suitable for ELISpot, FluoroSpot, and flow. Dissolve in DMSO then PBS; use at 2 µg/ml of each peptide. Store at -20 °C or below. Catalog 3617-1.
Catalog No. NC1925864 Supplier Mabtech Inc Supplier No. 36171
PepPool: CEFTA (CD4), human
Peptide pool for use as a positive control for human CD4 T-cell stimulation in ELISpot, FluoroSpot, and flow cytometry. Consists of 35 MHC class II-restricted peptides from human CMV, EBV, influenza virus, tetanus toxin, and adenovirus 5. Designed for broad HLA coverage. Synthetic peptides >95% pure. Induces IFN-γ, IL-2, IL-5, and IL-13 from antigen-specific CD4 T cells. For research use only. Not for diagnostic use.
Contents: One vial, lyophilized peptide pool, 35 peptides, 25 µg of each.
HLA coverage: HLA-DRB1*01, *03, *04, *07, *08, *11, *13, *15, *16, *53; HLA-DPw2; HLA-DPB1*04; HLA-DQw1; HLA-DQw3; HLA-DQB1*06.
Benefits: Validated positive control for CD4 responses; suitable for ELISpot, FluoroSpot, and flow. Dissolve in DMSO then PBS; use at 2 µg/ml of each peptide. Store at -20 °C or below. Catalog 3617-1.