Learn More
Medchemexpress LLC Ceftaroline 10mg | 189345-04-8 | 10MG
SDP
Catalog No. 5000243518
Encompass_Preferred
T-91825 (PPI-0903M) an N-phosphono-type cephalosporin is the active form of TAK-599 T-91825 is active against both gram-positive and gram-negative bacteria[1 [2
Catalog No. 50-002-43518 Supplier Medchemexpress LLC Supplier No. HY10504910MG
T-91825 (PPI-0903M) an N-phosphono-type cephalosporin is the active form of TAK-599 T-91825 is active against both gram-positive and gram-negative bacteria[1 [2