Learn More
Medchemexpress LLC Ceftaroline | 189345-04-8 | 96.9% ; 98.5% | 604.7 g/mol | C22H20N8O5S4 | 50 MG
SDP
Catalog No. 5000243690
Encompass_Preferred
Ceftaroline is an N-phosphono-type cephalosporin antibiotic and the active form of the prodrug ceftaroline fosamil. It exhibits activity against both Gram-positive and Gram-negative bacteria and is supplied as a research reagent for antibacterial studies.
  • Active metabolite of the prodrug ceftaroline fosamil.
  • Broad-spectrum activity against Gram-positive and Gram-negative bacteria.
  • Useful as a research reagent for antibacterial and microbiology studies.
  • Supplied in multiple package sizes for experimental needs.
  • High reported purity suitable for analytical applications.
Catalog No. 50-002-43690 Supplier Medchemexpress LLC Supplier No. HY10504950MG
Ceftaroline is an N-phosphono-type cephalosporin antibiotic and the active form of the prodrug ceftaroline fosamil. It exhibits activity against both Gram-positive and Gram-negative bacteria and is supplied as a research reagent for antibacterial studies.
  • Active metabolite of the prodrug ceftaroline fosamil.
  • Broad-spectrum activity against Gram-positive and Gram-negative bacteria.
  • Useful as a research reagent for antibacterial and microbiology studies.
  • Supplied in multiple package sizes for experimental needs.
  • High reported purity suitable for analytical applications.