Learn More
Medchemexpress LLC Ceftaroline | 189345-04-8 | 98.5% | 604.70 | C22H20N8O5S4 | 5 MG
SDP
Catalog No. 5000243113
Encompass_Preferred
Ceftaroline is an N-phosphono-type cephalosporin, the active metabolite form used in research to evaluate antibacterial activity against gram-positive and gram-negative organisms. It is supplied as a white to yellow solid for laboratory use and is suitable for in vitro and preclinical assays.
  • Antibacterial cephalosporin active metabolite.
  • Suitable for in vitro and preclinical research.
  • Supplied as a solid, white to yellow appearance.
  • Available in small vial sizes for assay work.
  • High reported purity (98.5%).
  • Molecular weight 604.70.
Catalog No. 50-002-43113 Supplier Medchemexpress LLC Supplier No. HY1050495MG
Ceftaroline is an N-phosphono-type cephalosporin, the active metabolite form used in research to evaluate antibacterial activity against gram-positive and gram-negative organisms. It is supplied as a white to yellow solid for laboratory use and is suitable for in vitro and preclinical assays.
  • Antibacterial cephalosporin active metabolite.
  • Suitable for in vitro and preclinical research.
  • Supplied as a solid, white to yellow appearance.
  • Available in small vial sizes for assay work.
  • High reported purity (98.5%).
  • Molecular weight 604.70.