Learn More
Medchemexpress LLC Ceftaroline | 189345-04-8 | 98.5% | 604.70 g·mol⁻¹ | C22H20N8O5S4 | 25 MG
SDP
Catalog No. 5000243829
Encompass_Preferred
Ceftaroline (T-91825) is an N-phosphono-type cephalosporin and the active metabolite of the prodrug ceftaroline fosamil. It demonstrates activity against a broad spectrum of gram-positive and gram-negative bacteria and is supplied as a high-purity research reagent for antibacterial studies.
  • High purity appropriate for analytical and in vitro biological assays.
  • Active metabolite of ceftaroline fosamil with established antibacterial profile.
  • Molecular formula C22H20N8O5S4; molecular weight 604.70 g·mol⁻¹.
  • Suitable for research on gram-positive and gram-negative pathogens.
  • Supplied as a small-quantity research pack for assay development and characterization.
Catalog No. 50-002-43829 Supplier Medchemexpress LLC Supplier No. HY10504925MG
Ceftaroline (T-91825) is an N-phosphono-type cephalosporin and the active metabolite of the prodrug ceftaroline fosamil. It demonstrates activity against a broad spectrum of gram-positive and gram-negative bacteria and is supplied as a high-purity research reagent for antibacterial studies.
  • High purity appropriate for analytical and in vitro biological assays.
  • Active metabolite of ceftaroline fosamil with established antibacterial profile.
  • Molecular formula C22H20N8O5S4; molecular weight 604.70 g·mol⁻¹.
  • Suitable for research on gram-positive and gram-negative pathogens.
  • Supplied as a small-quantity research pack for assay development and characterization.