Learn More
Medchemexpress LLC HY-14737 10mg , Ceftaroline fosamil CAS:400827-46-5 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1473710MG
Encompass_Preferred
HY-14737 10mg Ceftaroline fosamil CAS: 400827-46-5 Ceftaroline fosamil (TAK-599), a cephalosporin derivative, is an N-phosphono prodrug of anti-methicillin-resistant Staphylococcus aureus (MRSA) T-91825. Ceftaroline fosamil can be used for the research of MRSA infection. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-0879