Learn More
Medchemexpress LLC Ceftazidime-d5 (GR20263-d5) | 93.7% | 1 MG
SDP
Catalog No. 5000406744
Encompass_Preferred
Ceftazidime-d5 (GR20263-d5) is a deuterium-labeled antibiotic, specifically a version of Ceftazidime. Ceftazidime exhibits broad-spectrum activity against Gram-positive and Gram-negative aerobic bacteria, including Enterobacteriaceae (even β-lactamase-positive strains), and resists hydrolysis by most β-lactamases. Deuterium-labeled compounds, such as Ceftazidime-d5, are used as tracers for quantitative analysis during drug development, impacting pharmacokinetic and metabolic profiles.
  • Deuterium-labeled version of Ceftazidime
  • Exhibits broad-spectrum antibiotic activity
  • Effective against Gram-positive and Gram-negative aerobic bacteria
  • Active against Enterobacteriaceae, including β-lactamase-positive strains
  • Resistant to hydrolysis by most β-lactamases
  • Useful as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-06744 Supplier Medchemexpress LLC Supplier No. HYB0593S1MG
Ceftazidime-d5 (GR20263-d5) is a deuterium-labeled antibiotic, specifically a version of Ceftazidime. Ceftazidime exhibits broad-spectrum activity against Gram-positive and Gram-negative aerobic bacteria, including Enterobacteriaceae (even β-lactamase-positive strains), and resists hydrolysis by most β-lactamases. Deuterium-labeled compounds, such as Ceftazidime-d5, are used as tracers for quantitative analysis during drug development, impacting pharmacokinetic and metabolic profiles.
  • Deuterium-labeled version of Ceftazidime
  • Exhibits broad-spectrum antibiotic activity
  • Effective against Gram-positive and Gram-negative aerobic bacteria
  • Active against Enterobacteriaceae, including β-lactamase-positive strains
  • Resistant to hydrolysis by most β-lactamases
  • Useful as a tracer
  • Serves as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS