Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceftazidime Pentahydrate, G-Biosciences
SCI_ED

Catalog No. S910541
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S910541 1 g
S877438 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S910541 Supplier G-Biosciences Supplier No. RC947
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ceftazidime pentahydrate is a third generation cephalosporin antibiotic. It has been used to study the penicillin binding proteins, especially PBP3. It has also been used as a selection tool for mutant colonies.

Ceftazidime targets gram-negative bacteria, including Pseudomonas aeruginosa and is less effective against gram-positive cocci.

Chemical Identifiers

Molecular Weight (g/mol) 636.65

Specifications

Physical Form Powder
Quantity 1 g
Product Line Gibco
Boiling Point 319.08°C
Percent Purity >98%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.