Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Cefteram pivoxil | 82547-81-7 | 98.2% | 593.6 g/mol | C22H27N9O7S2 | 10 MG
SDP

Catalog No. 5000241007
Encompass_Preferred

Cefteram pivoxil is an orally active cephalosporin antibiotic prodrug of cefteram, used in research on bacterial infections. Provided as a purified chemical standard for laboratory studies, it is intended for research use only.

  • Orally active cephalosporin antibiotic prodrug.
  • Suitable for in vitro and in vivo bacterial infection studies.
  • High purity (98.18%) for analytical and experimental use.
  • Available in small-quantity packs for research flexibility.
  • Molecular weight 593.6 g/mol; formula C22H27N9O7S2.
  • CAS number 82547-81-7 for unambiguous identification.

Catalog No. 50-002-41007 Supplier Medchemexpress LLC Supplier No. HY10657110MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Cefteram pivoxil is an orally active cephalosporin antibiotic prodrug of cefteram, used in research on bacterial infections. Provided as a purified chemical standard for laboratory studies, it is intended for research use only.

  • Orally active cephalosporin antibiotic prodrug.
  • Suitable for in vitro and in vivo bacterial infection studies.
  • High purity (98.18%) for analytical and experimental use.
  • Available in small-quantity packs for research flexibility.
  • Molecular weight 593.6 g/mol; formula C22H27N9O7S2.
  • CAS number 82547-81-7 for unambiguous identification.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.