Learn More
Medchemexpress LLC Cefteram pivoxil | 82547-81-7 | 98.2% | 593.64 g·mol⁻¹ | C22H27N9O7S2 | 5 MG
SDP
Catalog No. 5000240055
Encompass_Preferred
Cefteram pivoxil is an orally active cephalosporin antibiotic prodrug used in research to study bacterial infections. It is supplied as a solid with formula C22H27N9O7S2 and a molecular weight of 593.64 g·mol⁻¹.
  • Orally active cephalosporin prodrug.
  • Third-generation broad-spectrum antibacterial profile.
  • High purity, 98.18% (supplier reported).
  • Solid form suitable for storage at -20°C or 4°C as recommended.
  • Small research-scale quantity (5 mg) for assay development.
Catalog No. 50-002-40055 Supplier Medchemexpress LLC Supplier No. HY1065715MG
Cefteram pivoxil is an orally active cephalosporin antibiotic prodrug used in research to study bacterial infections. It is supplied as a solid with formula C22H27N9O7S2 and a molecular weight of 593.64 g·mol⁻¹.
  • Orally active cephalosporin prodrug.
  • Third-generation broad-spectrum antibacterial profile.
  • High purity, 98.18% (supplier reported).
  • Solid form suitable for storage at -20°C or 4°C as recommended.
  • Small research-scale quantity (5 mg) for assay development.