Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cefteram Pivoxil, G-Biosciences
SCI_ED

Catalog No. S976226
Change view
Click to view available options
Quantity:
10 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S976226 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S976226 Supplier G-Biosciences Supplier No. RC949
Add to Cart
Add to Cart

Cefteram pivoxil is a semi-synthetic third generation cephalosporin antibacterial that is a prodrug ester of cefteram. It has been seen to express greater bactericidal predominantly against gram-negative bacteria.

Cefteram pivoxil has also induced IgE antibody immune responses during in vivo immunology studies.

Chemical Identifiers

CAS 82547-81-7
Molecular Weight (g/mol) 593.63 g/mol

Specifications

Technique HPLC, Spectrometry, UV/Vis, Gas Chromatography, GC-MS
Certifications/Compliance ISO, ISO 9001, REACH, NIST, GHS/OSHA
Type Drug Standard
Element Multi-element
Industry Type Biotech
Quantity 10 mg
Percent Purity >98%
Grade Antibiotic
Chemical Name or Material Cefteram pivoxil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.