Learn More
Medchemexpress LLC Ceftezole sodium | 41136-22-5 | 1 ML
SDP
Catalog No. 5000419766
Encompass_Preferred
Ceftezole sodium is a broad-spectrum cephem antibiotic that is effective against many species of gram-positive and gram-negative bacteria. It also acts as an alpha-glucosidase inhibitor with in vivo anti-diabetic activity.
  • Broad-spectrum cephem antibiotic
  • Effective against many species of gram-positive and gram-negative bacteria
  • Acts as an alpha-glucosidase inhibitor
  • Has in vivo anti-diabetic activity
  • Purity of 98.75%
  • White to off-white solid appearance
Catalog No. 50-004-19766 Supplier Medchemexpress LLC Supplier No. HYN709610MM/1ML
Ceftezole sodium is a broad-spectrum cephem antibiotic that is effective against many species of gram-positive and gram-negative bacteria. It also acts as an alpha-glucosidase inhibitor with in vivo anti-diabetic activity.
  • Broad-spectrum cephem antibiotic
  • Effective against many species of gram-positive and gram-negative bacteria
  • Acts as an alpha-glucosidase inhibitor
  • Has in vivo anti-diabetic activity
  • Purity of 98.75%
  • White to off-white solid appearance