Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceftibuten, G-Biosciences
SCI_ED

Catalog No. S932433
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S932433 10 mg
S666686 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S932433 Supplier G-Biosciences Supplier No. RC950
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ceftibuten is a third generation cephalosporin antibiotic targeting a broad-spectrum of gram-positive and gram-negative bacteria.

It works by binding to proteins in the bacterial cell wall and inhibiting its synthesis, leading to cell lysis and death.

Chemical Identifiers

Molecular Weight (g/mol) 410.42

Specifications

Physical Form Powder
Quantity 10 mg
Product Line Gibco
Boiling Point 966.4°C
Percent Purity >95%
Grade Molecular Biology
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.