Learn More
Cayman Chemical Ceftibuten (hydrate), Purity: ≥98%, CAS: 118081-34-8, 50 mg
SDP
Catalog No. 502932472
Encompass_Preferred
A broad spectrum cephalosporin antibiotic; inhibits the growth of B. catarrhalis, H. influenzae, pathogenic Neisseria, Streptococcus, penicillin-susceptible S. pneumoniae, and eleven Enterobacteriaceae strains (MICs = 0.25, ≤0.06, ≤0.06, 0.5-1, 4, and ≤0.06-0.5 μg/ml, respectively); active against E. coli expressing β-lactamase type I, III, and V and K. oxytoca expressing β-lactamase type IV (MICs = 8, 0.25, 0.5, and ≤0.06 μg/ml, respectively) as well as a panel of eight bacterial stains expressing plasmid-encoded extended spectrum β-lactamases, including E. coli CTX-M and K. pneumoniae SHV-2 (MICs = 1 and 0.25 μg/ml, respectively); reduces the number of mice killed within 6 days of a K. pneumoniae, E. coli, S. pneumoniae, or S. aureus infection at 0.125, 2, 1,024, and >512 mg/kg, respectively
Catalog No. 50-293-2472 Supplier Cayman Chemical Supplier No. 2533450 MG
A broad spectrum cephalosporin antibiotic; inhibits the growth of B. catarrhalis, H. influenzae, pathogenic Neisseria, Streptococcus, penicillin-susceptible S. pneumoniae, and eleven Enterobacteriaceae strains (MICs = 0.25, ≤0.06, ≤0.06, 0.5-1, 4, and ≤0.06-0.5 μg/ml, respectively); active against E. coli expressing β-lactamase type I, III, and V and K. oxytoca expressing β-lactamase type IV (MICs = 8, 0.25, 0.5, and ≤0.06 μg/ml, respectively) as well as a panel of eight bacterial stains expressing plasmid-encoded extended spectrum β-lactamases, including E. coli CTX-M and K. pneumoniae SHV-2 (MICs = 1 and 0.25 μg/ml, respectively); reduces the number of mice killed within 6 days of a K. pneumoniae, E. coli, S. pneumoniae, or S. aureus infection at 0.125, 2, 1,024, and >512 mg/kg, respectively