Learn More
Medchemexpress LLC Ceftiofur hydrochloride | 103980-44-5 | 98.4% | C19H18ClN5O7S3 | 1 ML
SDP
Catalog No. 5000432492
Encompass_Preferred
Ceftiofur hydrochloride is a potent cell wall synthesis inhibitor that targets bacterial penicillin-binding proteins (PBPs). It exerts bactericidal effects by inhibiting bacterial cell wall peptidoglycan synthesis, leading to cell lysis. Additionally, it possesses anti-inflammatory properties, inhibiting NF-κB and MAPKs, which reduces the secretion of pro-inflammatory cytokines such as TNF-α, IL-1β, and IL-6. This compound has been shown to improve survival rates in endotoxemia models.
  • Targets bacterial penicillin-binding proteins (PBPs)
  • Inhibits bacterial cell wall peptidoglycan synthesis
  • Induces bacterial cell lysis
  • Reduces secretion of pro-inflammatory cytokines (TNF-α, IL-1β, IL-6)
  • Inhibits phosphorylation of ERK, JNK, and p38
  • Blocks nuclear translocation of NF-κB p65
  • Improves survival in endotoxemia models
  • Cited in 4 scientific publications
Catalog No. 50-004-32492 Supplier Medchemexpress LLC Supplier No. HYB002610MM/1ML
Ceftiofur hydrochloride is a potent cell wall synthesis inhibitor that targets bacterial penicillin-binding proteins (PBPs). It exerts bactericidal effects by inhibiting bacterial cell wall peptidoglycan synthesis, leading to cell lysis. Additionally, it possesses anti-inflammatory properties, inhibiting NF-κB and MAPKs, which reduces the secretion of pro-inflammatory cytokines such as TNF-α, IL-1β, and IL-6. This compound has been shown to improve survival rates in endotoxemia models.
  • Targets bacterial penicillin-binding proteins (PBPs)
  • Inhibits bacterial cell wall peptidoglycan synthesis
  • Induces bacterial cell lysis
  • Reduces secretion of pro-inflammatory cytokines (TNF-α, IL-1β, IL-6)
  • Inhibits phosphorylation of ERK, JNK, and p38
  • Blocks nuclear translocation of NF-κB p65
  • Improves survival in endotoxemia models
  • Cited in 4 scientific publications