Learn More
Medchemexpress LLC Ceftiofur hydrochloride | 103980-44-5 | MFCD04038020 | 98.43% | C19H18ClN5O7S3 | 100 MG
SDP
Catalog No. 5000432051
Encompass_Preferred
Ceftiofur hydrochloride is an inhibitor of bacterial cell wall synthesis that targets bacterial penicillin-binding proteins (PBPs). It exhibits anti-inflammatory effects in endotoxemia. Its bactericidal action stems from inhibiting the synthesis of bacterial cell wall peptidoglycan, leading to bacterial cell lysis. Additionally, Ceftiofur hydrochloride can inhibit the activation of NF-κB and MAPKs, which consequently reduces the secretion of pro-inflammatory cytokines such as TNF-α, IL-1β, and IL-6.
  • Targets bacterial penicillin-binding proteins (PBPs).
  • Exhibits bactericidal effects by inhibiting bacterial cell wall peptidoglycan synthesis.
  • Leads to bacterial cell lysis.
  • Possesses anti-inflammatory properties by inhibiting NF-κB and MAPK activation.
  • Reduces the secretion of pro-inflammatory cytokines (TNF-α, IL-1β, IL-6).
Catalog No. 50-004-32051 Supplier Medchemexpress LLC Supplier No. HYB0026100MG
Ceftiofur hydrochloride is an inhibitor of bacterial cell wall synthesis that targets bacterial penicillin-binding proteins (PBPs). It exhibits anti-inflammatory effects in endotoxemia. Its bactericidal action stems from inhibiting the synthesis of bacterial cell wall peptidoglycan, leading to bacterial cell lysis. Additionally, Ceftiofur hydrochloride can inhibit the activation of NF-κB and MAPKs, which consequently reduces the secretion of pro-inflammatory cytokines such as TNF-α, IL-1β, and IL-6.
  • Targets bacterial penicillin-binding proteins (PBPs).
  • Exhibits bactericidal effects by inhibiting bacterial cell wall peptidoglycan synthesis.
  • Leads to bacterial cell lysis.
  • Possesses anti-inflammatory properties by inhibiting NF-κB and MAPK activation.
  • Reduces the secretion of pro-inflammatory cytokines (TNF-α, IL-1β, IL-6).