Learn More
Medchemexpress LLC Ceftiofur (sodium) | 104010-37-9 | 98.0% | 1 ML
SDP
Supplier:  Medchemexpress LLC HYB089810MM/1ML
Encompass_Preferred
Ceftiofur sodium is a cell wall synthesis inhibitor that targets bacterial penicillin-binding proteins (PBPs) and demonstrates anti-inflammatory effects in endotoxemia. It acts by inhibiting the synthesis of bacterial cell wall peptidoglycan, which leads to bacterial cell lysis. Additionally, it inhibits the activation of NF-κB and MAPKs, thereby reducing the secretion of pro-inflammatory cytokines like TNF-α, IL-1β, and IL-6.
  • Targets bacterial penicillin-binding proteins
  • Inhibits bacterial cell wall synthesis
  • Has anti-inflammatory effects in endotoxemia
  • Reduces pro-inflammatory cytokine secretion (TNF-α, IL-1β, IL-6)
  • Inhibits NF-κB and MAPK activation
Catalog No. 50-004-09261