Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ceftiofur Sodium, G-Biosciences
SCI_ED

Catalog No. S910518
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S910518 1 g
S791027 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. S910518 Supplier G-Biosciences Supplier No. RC952
Add to Cart
Add to Cart

Ceftiofur sodium is a third generation cephalosporin antibiotic effective against gram-positive and gram-negative bacteria.

Although ceftiofur is resistant to Beta-lactamase degradation, there are several strains of E. coli known to be unaffected by this antibiotic.

Chemical Identifiers

Molecular Weight (g/mol) 545.54 g/mol

Specifications

Quantity 1 g
Percent Purity >95%
Grade Molecular Biology
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.