Learn More
Cayman Chemical Ceftiofur (hydrochloride), Purity: ≥98%, CAS: 103980-44-5, 10 g
SDP
Catalog No. 502939760
Encompass_Preferred
A broad-spectrum cephalosporin antibiotic; active against porcine isolates of A. pleuropneumoniae, P. multocida, S. typhimurium, S. cholerae-suis, E. coli, S. suis, and β-hemolytic streptococci (MIC90s = ≤2 μg/ml); exhibits curative rates of 43, 88, and 100% in a lactating bovine model of intramammary S. uberis infection when administered via intramammary infusion at a dose of 125 mg per infected quarter for 2, 5, and 8 days, respectively
Catalog No. 50-293-9760 Supplier Cayman Chemical Supplier No. 2730110 G
A broad-spectrum cephalosporin antibiotic; active against porcine isolates of A. pleuropneumoniae, P. multocida, S. typhimurium, S. cholerae-suis, E. coli, S. suis, and β-hemolytic streptococci (MIC90s = ≤2 μg/ml); exhibits curative rates of 43, 88, and 100% in a lactating bovine model of intramammary S. uberis infection when administered via intramammary infusion at a dose of 125 mg per infected quarter for 2, 5, and 8 days, respectively